EMAIL THIS PAGE TO A FRIEND
Stem Cells and Development
Bovine posterior limbus: an evaluation of an alternative source for corneal endothelial and trabecular meshwork stem/progenitor cells.
PMID 25323922