EMAIL THIS PAGE TO A FRIEND
Journal of neurosurgery. Pediatrics
Ventriculoperitoneal shunt infection with Mycobacterium fortuitum: a rare offending organism.
PMID 25325413
Abstract
Postsurgical infection is one of the greatest potential morbidities of ventriculoperitoneal shunt surgery. The majority of infections can be linked to contamination with skin flora at the time of surgery, a phenomenon that has been well described. However, there is a paucity of literature regarding infection with nontuberculous mycobacteria. The authors report a case of postoperative ventriculoperitoneal shunt infection with Mycobacterium fortuitum and review the available neurosurgical literature and treatment strategies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2487 Cotrimoxazole, Ready Made Solution, 100 mg/mL in DMSO
C14H18N4O3 · 5 C10H11N3O3S