EMAIL THIS PAGE TO A FRIEND
Medical Oncology (Northwood, London, England)
Long noncoding RNA GAS5 affects cell proliferation and predicts a poor prognosis in patients with colorectal cancer.
PMID 25326054