EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Seizure control by derivatives of medium chain fatty acids associated with the ketogenic diet show novel branching-point structure for enhanced potency.
PMID 25326131
Abstract
The medium chain triglyceride (MCT) ketogenic diet is a major treatment of drug-resistant epilepsy but is problematic, particularly in adults, because of poor tolerability. Branched derivatives of octanoic acid (OA), a medium chain fat provided in the diet have been suggested as potential new treatments for drug-resistant epilepsy, but the structural basis of this functionality has not been determined. Here we investigate structural variants of branched medium chain fatty acids as new seizure-control treatments. We initially employ a series of methyl-branched OA derivatives, and using the GABAA receptor antagonist pentylenetetrazol to induce seizure-like activity in rat hippocampal slices, we show a strong, branch-point-specific activity that improves upon the related epilepsy treatment valproic acid. Using low magnesium conditions to induce glutamate excitotoxicity in rat primary hippocampal neuronal cultures for the assessment of neuroprotection, we also show a structural dependence identical to that for seizure control, suggesting a related mechanism of action for these compounds in both seizure control and neuroprotection. In contrast, the effect of these compounds on histone deacetylase (HDAC) inhibition, associated with teratogenicity, shows no correlation with therapeutic efficacy. Furthermore, small structural modifications of the starting compounds provide active compounds without HDAC inhibitory effects. Finally, using multiple in vivo seizure models, we identify potent lead candidates for the treatment of epilepsy. This study therefore identifies a novel family of fatty acids, related to the MCT ketogenic diet, that show promise as new treatments for epilepsy control and possibly other MCT ketogenic diet-responding conditions, such as Alzheimer disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0426000
Caprylic acid, European Pharmacopoeia (EP) Reference Standard
C8H16O2
PHR1202
Caprylic Acid (Octanoic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
C8H16O2
13112
Magnesium, powder, ≥99%
Mg
13103
Magnesium, ribbon, ≥99.5% Mg basis
Mg
474754
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Mg
63035
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Mg
200905
Magnesium, turnings, reagent grade, 98%
Mg
254118
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Mg
254126
Magnesium, 20-230 mesh, reagent grade, 98%
Mg
403148
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Mg
266302
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Mg
465666
Magnesium, ReagentPlus®, powder, −325 mesh, 99.5% trace metals basis
Mg
63040
Magnesium, grit, ≥99.0% (KT)
Mg
253987
Magnesium, ReagentPlus®, powder, −50 mesh, ≥99%
Mg
299405
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Mg
734365
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Mg
GF58454983
Magnesium, wire reel, 1m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF96495609
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Mg
735779
Magnesium, ingot, ≥99% trace metals basis
Mg
GF04987547
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF72273157
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF34143803
Magnesium, wire reel, 1m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF23637371
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF30456053
Magnesium, wire reel, 1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF83829178
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF28467636
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF54671924
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Mg
723320
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Mg
GF18270637
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF23443646
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%
Mg