EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
IKKβ is an IRF5 kinase that instigates inflammation.
PMID 25326420
Abstract
The transcription factor interferon regulatory factor 5 (IRF5) is essential for the induction of inflammatory cytokines, but the mechanism by which IRF5 is activated is not well understood. Here we present evidence that the kinase IKKβ phosphorylates and activates IRF5 in response to stimulation in several inflammatory pathways, including those emanated from Toll-like receptors and retinoic acid-inducible gene I-like receptors. IKKβ phosphorylates mouse IRF5 at specific residues, including serine 445 (S446 in human IRF5 isoform 1), as evidenced by mass spectrometry analysis and detection with a phosphospecific antibody. Recombinant IKKβ phosphorylated IRF5 at Ser-445 in vitro, and a point mutation of this serine abolished IRF5 activation and cytokine production. Depletion or pharmacologic inhibition of IKKβ prevented IRF5 phosphorylation. These results indicate that IKKβ is an IRF5 kinase that instigates inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN1027 CompoZr® Knockout ZFN Kit, human IKBKB (NM_001556)