EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
High-performance liquid chromatographic enantioseparation of cationic 1,2,3,4-tetrahydroisoquinoline analogs on Cinchona alkaloid-based zwitterionic chiral stationary phases.
PMID 25326889