EMAIL THIS PAGE TO A FRIEND
Nature
Reductive dehalogenase structure suggests a mechanism for B12-dependent dehalogenation.
PMID 25327251
Abstract
Organohalide chemistry underpins many industrial and agricultural processes, and a large proportion of environmental pollutants are organohalides. Nevertheless, organohalide chemistry is not exclusively of anthropogenic origin, with natural abiotic and biological processes contributing to the global halide cycle. Reductive dehalogenases are responsible for biological dehalogenation in organohalide respiring bacteria, with substrates including polychlorinated biphenyls or dioxins. Reductive dehalogenases form a distinct subfamily of cobalamin (B12)-dependent enzymes that are usually membrane associated and oxygen sensitive, hindering detailed studies. Here we report the characterization of a soluble, oxygen-tolerant reductive dehalogenase and, by combining structure determination with EPR (electron paramagnetic resonance) spectroscopy and simulation, show that a direct interaction between the cobalamin cobalt and the substrate halogen underpins catalysis. In contrast to the carbon-cobalt bond chemistry catalysed by the other cobalamin-dependent subfamilies, we propose that reductive dehalogenases achieve reduction of the organohalide substrate via halogen-cobalt bond formation. This presents a new model in both organohalide and cobalamin (bio)chemistry that will guide future exploitation of these enzymes in bioremediation or biocatalysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
252018
2,6-Dibromophenol, 99%
C6H4Br2O
266639
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Co
266647
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Co
697745
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Co
203076
Cobalt, granular, 99.99% trace metals basis
Co
356867
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Co
266655
Cobalt, pieces, 99.5% trace metals basis
Co
356891
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Co
398810
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Co
GF17608091
Cobalt, wire reel, 2m, diameter 0.05mm, hard, 99.9%
Co
266671
Cobalt, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Co
GF48813723
Cobalt, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Co
GF55348140
Cobalt, foil, 25mm disks, thickness 0.05mm, annealed, 99.9%
Co
GF44764993
Cobalt, foil, 25mm disks, thickness 0.05mm, as rolled, 99.9%
Co
GF95811475
Cobalt, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%
Co
GF94594450
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
Co
GF41541476
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Co
GF45372315
Cobalt, foil, 4mm disks, thickness 0.003mm, 99.9%
Co
GF08560289
Cobalt, foil, 4mm disks, thickness 0.004mm, 99.9%
Co
GF89559343
Cobalt, foil, 4mm disks, thickness 0.005mm, 99.9%
Co
GF48392051
Cobalt, foil, 4mm disks, thickness 0.006mm, 99.9%
Co
GF55954873
Cobalt, foil, 4mm disks, thickness 0.007mm, 99.9%
Co
GF92926727
Cobalt, foil, 4mm disks, thickness 0.008mm, 99.9%
Co
GF70445246
Cobalt, foil, 4mm disks, thickness 0.009mm, 99.9%
Co
GF40089225
Cobalt, foil, 4mm disks, thickness 0.0125mm, 99.9%
Co
GF03600278
Cobalt, foil, 4mm disks, thickness 0.015mm, 99.9%
Co
GF78079630
Cobalt, foil, 4mm disks, thickness 0.01mm, 99.9%
Co
GF29762646
Cobalt, foil, 4mm disks, thickness 0.025mm, 99.9%
Co
GF97588300
Cobalt, foil, 4mm disks, thickness 0.02mm, 99.9%
Co
GF21186985
Cobalt, foil, 4mm disks, thickness 0.05mm, annealed, 99.9%
Co