EMAIL THIS PAGE TO A FRIEND
Nature Communications
Transparent and flexible low noise graphene electrodes for simultaneous electrophysiology and neuroimaging.
PMID 25327632
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14343
1(S),9(R)-(−)-Bicuculline methiodide, ≥95.0% (HPCE)
C21H20INO6