EMAIL THIS PAGE TO A FRIEND
Zahn-, Mund-, und Kieferheilkunde mit Zentralblatt
[The porosity, microhardness, roughness and internal stresses of PMMA base materials in relation to their isolation. 1. Porosity and microhardness].
PMID 2532829
Abstract
Selected material properties of hot and cold polymerized denture basic materials (polymethylmethacrylate) were investigated after different methods of isolation: alginate, tin foil and silicone. Both tin foil and silicone isolation the handling of which is much easier result in a decreased portion of porosities and an increased microhardness in the PMMA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF32793509
Tin, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Sn
GF27720019
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Sn
GF28603212
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 98.8%
Sn
GF63204106
Tin, foil, 4mm disks, thickness 1.0mm, as rolled, 100%
Sn
GF11466839
Tin, foil, 50x50mm, thickness 3.0mm, as rolled, 99.99+%
Sn
GF43514535
Tin, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Sn
GF67327375
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF16685219
Tin, foil, 150x150mm, thickness 0.20mm, as rolled, 98.8%
Sn
GF26583194
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF77851323
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 98.8%
Sn
GF39022815
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 98.8%
Sn
GF89426807
Tin, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Sn
GF57824095
Tin, foil, 15mm disks, thickness 0.015mm, 99.75%
Sn
GF02405360
Tin, foil, 2m coil, thickness 0.009mm, 97.4%
Sn
GF99188252
Tin, foil, 50x50mm, thickness 1.0mm, as rolled, 99.999%
Sn
GF83290440
Tin, foil, 50x50mm, thickness 2.0mm, as rolled, 99.99+%
Sn
GF02196222
Tin, foil, 0.5m coil, thickness 0.01mm, 97.4%
Sn
GF64405762
Tin, foil, 100x100mm, thickness 2.0mm, as rolled, 98.8%
Sn
GF07913088
Tin, foil, 8mm disks, thickness 0.06mm, as rolled, 99.99+%
Sn
GF21766068
Tin, foil, 100x100mm, thickness 0.15mm, as rolled, 99.99+%
Sn
GF48212669
Tin, foil, 100x100mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF96022852
Tin, foil, 2m coil, thickness 0.125mm, as rolled, 98.8%
Sn
GF59159223
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99+%
Sn
GF70491731
Tin, foil, 25x25mm, thickness 3.0mm, as rolled, 99.99+%
Sn
GF30417566
Tin, foil, 0.5m coil, thickness 0.0125mm, 97.4%
Sn
GF76227138
Tin, foil, 0.5m coil, thickness 0.1mm, as rolled, 99.95%
Sn
GF11564535
Tin, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Sn
GF08666321
Tin, foil, 25x25mm, thickness 2.0mm, as rolled, 99.99+%
Sn
GF52715415
Tin, foil, 50mm disks, thickness 0.5mm, as rolled, 98.8%
Sn
GF93985701
Tin, foil, 50x50mm, thickness 0.125mm, as rolled, 98.8%
Sn