EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Thiazolothiazole-linked porous organic polymers.
PMID 25329834
Abstract
Thiazolothiazole-linked porous organic polymers have been synthesized from a facile catalyst-free condensation reaction between aldehydes and dithiooxamide under solvothermal conditions. The resultant porous frameworks exhibit a highly selective uptake of CO2 over N2 under ambient conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
379387
Dithiooxamide, 97%
C2H4N2S2