EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Flexible low-voltage organic complementary circuits: finding the optimum combination of semiconductors and monolayer gate dielectrics.
PMID 25330764
Abstract
Low-voltage p-channel and n-channel organic transistors with channel lengths down to 0.5 μm using four small-molecule semiconductors and ultra-thin dielectrics based on two different phosphonic acid monolayers are fabricated on plastic substrates and studied in terms of effective mobility, intrinsic mobility and contact resistance. For the optimum materials combination, flexible complementary circuits have signal delays of 3.1 μs at 5 V.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
767638
Dinaphtho[2,3-b:2′,3′-f]thieno[3,2-b]thiophene, sublimed grade, 99%
C22H12S2