EMAIL THIS PAGE TO A FRIEND
Aaps Pharmscitech
Determining the polymer threshold amount for achieving robust drug release from HPMC and HPC matrix tablets containing a high-dose BCS class I model drug: in vitro and in vivo studies.
PMID 25331194