EMAIL THIS PAGE TO A FRIEND
European journal of nuclear medicine and molecular imaging
Uptake and tracer kinetics of O-(2-(18)F-fluoroethyl)-L-tyrosine in meningiomas: preliminary results.
PMID 25331459
Abstract
O-(2-[(18)F]Fluoroethyl)-L-tyrosine ((18)F-FET) is a well-established PET tracer for the imaging of cerebral gliomas, but little is known about (18)F-FET uptake in meningiomas. The aim of this study was to explore (18)F-FET kinetics and tumour-to-background contrast in meningiomas of various histologies. A group of 24 patients with suspected cerebral meningioma on MRI/CT had an additional dynamic (18)F-FET PET scan prior to surgery. Time-activity curves (TAC) of (18)F-FET uptake in the tumours and tumour-to-brain ratios (TBR) for early (3xa0-xa014xa0min after injection) and late (18)F-FET uptake (20xa0-xa040xa0min after injection) were analysed and compared with histological subtypes and WHO grade. (18)F-FET uptake in critical structures in the skull base was also evaluated in terms of tumour-to-tissue (T/Tis) ratio. TBR of (18)F-FET uptake in meningiomas was significantly higher in the early phase than in the late phase (3.5 ± 0.8 vs. 2.2 ± 0.3; P < 0.001). The difference in TBR between low-grade meningiomas (WHO grade I, 18 patients) and high-grade meningiomas (WHO grade II or III, 6 patients) was significant in the late phase of (18)F-FET uptake (2.1 ± 0.2 vs. 2.5 ± 0.2, P = 0.003) while there was no significant difference in the early phase. ROC analysis showed that TBR of (18)F-FET uptake in the late phase had significant power to differentiate low-grade from high-grade meningiomas (AUC 0.87 ± 0.18, sensitivity 83xa0%, specificity 83xa0%, optimal cut-off 2.3; P < 0.01). Evaluation of TAC yielded three different curve patterns of (18)F-FET PET uptake. Combination of TBR (cut-off value 2.3) and TAC pattern slightly improved the differentiation of high-grade from low-grade meningiomas (accuracy 92xa0%; P = 0.001). Analysis of background radioactivity in the skull base indicated that (18)F-FET uptake may be helpful in distinguishing meningioma tissue in the late phase. T/Tis ratios were >1.2 in all patients for the periorbita, sphenoidal sinus, pituitary gland, tentorium, bone and brain, in more than 90xa0% of patients for the mucosa and dura, but in only 63xa0% of patients for the cavernous sinus. (18)F-FET PET may provide additional information for noninvasive grading of meningiomas and possibly for the discrimination of tumour in critical areas of the skull base. A further evaluation of (18)F-FET PET in meningiomas appears to be justified.