EMAIL THIS PAGE TO A FRIEND
Infection and Immunity
Accumulation-associated protein enhances Staphylococcus epidermidis biofilm formation under dynamic conditions and is required for infection in a rat catheter model.
PMID 25332125
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04260
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
C6H7NO2