EMAIL THIS PAGE TO A FRIEND
Cell transplantation
Intra-arterial bone marrow mononuclear cell transplantation correlates with GM-CSF, PDGF-BB, and MMP-2 serum levels in stroke patients: results from a clinical trial.
PMID 25335778
Abstract
Bone marrow mononuclear cell (BM-MNC) intra-arterial transplantation improves recovery in experimental models of ischemic stroke through secretion of cytokines and growth factors (GFs), enhancing neoangiogenesis, and enhancing neuroplasticity. In this study, we tested whether BM-MNC transplantation in stroke patients induces changes in serum levels of cytokines and GFs. A phase I/II trial was conducted in middle cerebral artery (MCA) stroke patients with autologous intra-arterial BM-MNC transplantation between 5 and 9 days after stroke. Follow-up was done for up to 6 months. Eight cases and nine controls were included, and the serum levels of granulocyte-macrophage colony-stimulating factor (GM-CSF), platelet-derived growth factor-BB (PDGF-BB), β nerve growth factor (β-NGF), and matrix metalloproteinases 2 (MMP-2) and 9 (MMP-9) were measured before and 4, 8, and 90 days after transplantation. The correlation of these serum levels with dose of cells and clinical outcomes was studied. A total of 1.59 × 10(8) (±1.21 × 10(8)) BM-MNCs were injected in cases; of them 3.38 × 10(6) (±2.33 × 10(6)) were CD34(+) cells. There was a positive correlation between total BM-MNCs injected and levels of GM-CSF and PDGF-BB at 90 days after transplantation (r = 0.929, p = 0.001 and r = 0.714, p = 0.047, respectively), and a negative correlation between total CD34(+) cells injected and MMP-2 levels at 4 days after transplantation (r = -0.786, p = 0.036). Lower plasma levels of MMP-2 at 4 days and higher levels of PDGF-BB at 90 days were associated with better functional outcomes during follow-up (p = 0.019 and p = 0.037, respectively). When administered intra-arterially in subacute MCA stroke patients, BM-MNCs seem to induce changes in serum levels of GM-CSF, PDGF-BB, and MMP-2, even 3 months after transplantation, which could be associated with better functional outcomes. This manuscript is published as part of the International Association of Neurorestoratology (IANR) special issue of Cell Transplantation.