EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
O-GlcNAcase: promiscuous hexosaminidase or key regulator of O-GlcNAc signaling?
PMID 25336650
Abstract
O-GlcNAc signaling is regulated by an opposing pair of enzymes: O-GlcNAc transferase installs and O-GlcNAcase (OGA) removes the modification from proteins. The dynamics and regulation of this process are only beginning to be understood as the physiological functions of both enzymes are being probed using genetic and pharmacological approaches. This minireview charts the discovery and functional and structural analysis of OGA and summarizes the insights gained from recent studies using OGA inhibition, gene knock-out, and overexpression. We identify several areas of "known unknowns" that would benefit from future research, such as the enigmatic C-terminal domain of OGA.
Related Materials