EMAIL THIS PAGE TO A FRIEND
Journal of Experimental Botany
MONOPTEROS directly activates the auxin-inducible promoter of the Dof5.8 transcription factor gene in Arabidopsis thaliana leaf provascular cells.
PMID 25336688