EMAIL THIS PAGE TO A FRIEND
Journal of Clinical Microbiology
Multicentric comparative assessment of the bio-evolution Toxoplasma gondii detection kit with eight laboratory-developed PCR assays for molecular diagnosis of congenital toxoplasmosis.
PMID 25339393