PLoS ONE
Trypanosoma cruzi evades the protective role of interferon-gamma-signaling in parasite-infected cells.
PMID 25340519
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
447439
1-Methyl-L-tryptophan, 95%
C12H14N2O2