EMAIL THIS PAGE TO A FRIEND
Sexually Transmitted Infections
Drug use, sexual risk behaviour and sexually transmitted infections among swingers: a cross-sectional study in The Netherlands.
PMID 25342812