EMAIL THIS PAGE TO A FRIEND
Proteome Science
Impact of three commercial feed formulations on farmed gilthead sea bream (Sparus aurata, L.) metabolism as inferred from liver and blood serum proteomics.
PMID 25342931