EMAIL THIS PAGE TO A FRIEND
Mutation research. Genetic toxicology and environmental mutagenesis
Critical role of cellular glutathione homeostasis for trivalent inorganic arsenite-induced oxidative damage in human bronchial epithelial cells.
PMID 25344162
Abstract
Trivalent inorganic arsenic (iAs(3+)) is a powerful carcinogen that enhances the risk of lung cancer. Paradoxically, iAs(3+) also shows substantial efficacy in the treatment of lung tumors. However, the exact molecular mechanisms underlying iAs(3+)-induced toxicity and therapeutic effect in lung remain unclear. In this study, the effects of iAs(3+), sodium arsenite (NaAsO2) and arsenic trioxide (As2O3), on cell viability, apoptosis, genotoxicity and oxidative stress in cultured human bronchial epithelial cells were observed. Our results showed that NaAsO2 and As2O3 exposure could result in defects in cell proliferation and greatly enhance the level of oxidative damage. To clarify the critical role of glutathione (GSH) homeostasis in oxidative damage induced by iAs(3+), we further measured the content of GSH, ratio of GSH to GSSG, and the activities of GSH-related enzymes involved in the process of GSH synthesis, recycling and utilization. Our data demonstrated that NaAsO2 and As2O3 disrupted the balance of GSH homeostasis, and NaAsO2- and As2O3-induced oxidative damage was closely associated with the imbalance in GSH synthesis, recycling and utilization. To better understand the physiologic significance of Nrf2 in maintaining GSH-homeostasis, the expression level of Nrf2 was measured after iAs(3+) exposure. We found that the protein expression levels of Nrf2 were increased in both NaAsO2- and As2O3-treated cells. Collectively, our findings suggest that disturbed Nrf2-regulated GSH-homeostasis is associated with the oxidative damage triggered by iAs(3+), and loss of GSH homeostasis might implicate in both the pathogenesis of iAs(3+)-induced lung diseases and anticancer activity of iAs(3+).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2140
L-Glutathione oxidized disodium salt, suitable for cell culture, BioReagent
C20H30N6Na2O12S2