EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Inactivation of a GAL4-like transcription factor improves cell fitness and product yield in glycoengineered Pichia pastoris strains.
PMID 25344235