EMAIL THIS PAGE TO A FRIEND
American journal of reproductive immunology (New York, N.Y. : 1989)
Toll-like receptor-mediated responses by placental Hofbauer cells (HBCs): a potential pro-inflammatory role for fetal M2 macrophages.
PMID 25345551
Abstract
Microbial-driven responses in placenta are linked with adverse pregnancy outcomes. The role of Toll-like receptor (TLR) function in Hofbauer cells (HBCs) and fetal macrophages of the placental villous core remains understudied. Flow cytometry and immunohistochemistry (IHC) were used to establish the phenotype of HBCs. Regulation of cytokine secretion in response to treatment with TLR agonists and expression levels of TLRs and co-activators were compared in HBCs, placental fibroblasts (FIBs), and human umbilical vein endothelial cells (HUVECs) using ELISA and qPCR. Although flow cytometry and IHC revealed HBCs to be M2 (anti-inflammatory) macrophages, LPS and polyinosinic: polycytidylic acid [poly (I:C)] treatments markedly enhanced IL-6 secretion by HBCs, and expression of TLR-2, TLR-3, TLR-4, CD14, and MD-2 was the highest in HBCs. These results indicate that although HBCs are M2 macrophages, inflammatory responses are induced through TLR-3 and TLR-4 in this cell type, suggesting a role in microbial-driven placental/fetal inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50404
1,3-Propanediol, 98%
C3H8O2
1043003
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
C6H8O6
A1300000
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
C6H8O6
PHR1008
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
C6H8O6
95321
Hydrogen peroxide solution, ≥30%, for trace analysis
H2O2
16911
Hydrogen peroxide solution, for ultratrace analysis
H2O2
88597
Hydrogen peroxide solution, 3%, for microbiology
H2O2
95294
Hydrogen peroxide solution, tested according to Ph Eur
H2O2
95212
L-Ascorbic acid, tested according to Ph Eur
C6H8O6
57803
L-Ascorbic acid, certified reference material, TraceCERT®
C6H8O6
L1376
Linoleic acid, ≥99%
C18H32O2
L1012
Linoleic acid, liquid, BioReagent, suitable for cell culture
C18H32O2
62230
Linoleic acid, analytical standard
C18H32O2
62240
Linoleic acid, technical, 58-74% (GC)
C18H32O2
PHR1047
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
C6H6O
1524806
Phenol, United States Pharmacopeia (USP) Reference Standard
C6H6O
35952
Phenol, PESTANAL®, analytical standard
C6H6O
77608
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
C6H6O
46344
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C6H6O
48688
Phenol solution, certified reference material, 500 μg/mL in methanol
C6H6O
40063
Phenol solution, 5000 μg/mL in methanol, certified reference material
C6H6O
211176
Selenous acid, 98%
H2O3Se
229857
Selenous acid, 99.999% trace metals basis
H2O3Se