EMAIL THIS PAGE TO A FRIEND
Journal of Cardiovascular Electrophysiology
A TTX-sensitive resting Na+ permeability contributes to the catecholaminergic automatic activity in rat pulmonary vein.
PMID 25346483