EMAIL THIS PAGE TO A FRIEND
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society
Role of SUVmax and GLUT-1 Expression in Determining Tumor Aggressiveness in Patients With Clinical Stage I Endometrioid Endometrial Cancer.
PMID 25347093
Abstract
The aim of this study was to determine the role of (18)F-fluorodeoxyglucose positron emission tomography/computed tomography in estimating tumor aggressiveness in patients with clinical stage I endometrial cancer and the correlation between aggressiveness and expression of glucose transporter 1 (GLUT-1). F-fluorodeoxyglucose positron emission tomography/computed tomography was performed on 43 patients with clinical stage I endometrioid endometrial cancer. (18)F-fluorodeoxyglucose uptake was quantified by calculating the maximum standardized uptake value (SUV(max)) and GLUT-1 expression status based on immunohistochemistry. The mean (SD) SUV(max) of the primary tumor was 8.55 (5.04). The mean SUV(max) and GLUT-1 expression in stage IB and stage IC were significantly higher than that in stage IA (P = 0.001; P = 0.003). The mean (SD) SUV(max) was 6.81 (4.55) in grade 1, 10.92 (4.61) in grade 2, and 15.35 (1.34) in grade 3 (grade 1 vs grade 2 and 3; P = 0.005). The mean (SD) GLUT-1 expression was 1.17 (0.94) in grade 1, 2.00 (0.94) in grade 2, and 3.00 (0.00) in grade 3 (grade 1 vs grade 2 and 3; P = 0.017). Tumor aggressiveness, such as myometrial invasion or tumor grade, had a positive correlation with the SUV(max) and GLUT-1 expression in patients with clinical stage I endometrioid endometrial cancer.