EMAIL THIS PAGE TO A FRIEND
Free Radical Research
A new low molecular weight, MnII-containing scavenger of superoxide anion protects cardiac muscle cells from hypoxia/reoxygenation injury.
PMID 25348343