EMAIL THIS PAGE TO A FRIEND
Acta biochimica et biophysica Sinica
siRNA targeting stathmin inhibits invasion and enhances chemotherapy sensitivity of stem cells derived from glioma cell lines.
PMID 25348735