EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Molecularly imprinted solid-phase extraction for selective extraction of bisphenol analogues in beverages and canned food.
PMID 25350221