EMAIL THIS PAGE TO A FRIEND
Organic letters
Phenazinium salt-catalyzed aerobic oxidative amidation of aromatic aldehydes.
PMID 25350690
Abstract
Amides are prevalent in organic synthesis. Developing an efficient synthesis that avoids expensive oxidants and heating is highly desirable. Here the oxidative amidation of aromatic aldehydes is reported using an inexpensive metal-free visible light photocatalyst, phenazine ethosulfate, at low catalytic loading (1-2 mol %). The reaction proceeds at ambient temperature and uses air as the sole oxidant. The operationally easy procedure provides an economical, green, and mild alternative for the formation of amide bonds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4544
Phenazine ethosulfate, ≥95%
C16H18N2O4S