EMAIL THIS PAGE TO A FRIEND
Hypertension (Dallas, Tex. : 1979)
Prospects for droxidopa in neurogenic orthostatic hypotension.
PMID 25350983
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9446
L-DOPS, ≥98% (HPLC)
C9H11NO5