EMAIL THIS PAGE TO A FRIEND
Sao Paulo medical journal = Revista paulista de medicina
Greater expression of the human leukocyte antigen-G (HLA-G) and interleukin-17 (IL-17) in cervical intraepithelial neoplasia: analytical cross-sectional study.
PMID 25351636
Abstract
Impaired local cell immunity seems to contribute towards the pathogenesis and progression of cervical intraepithelial neoplasia (CIN), but the underlying molecular mechanisms promoting its progression remain unclear. Identification of new molecular markers for prognosis and diagnosis of early-stage CIN may aid in decreasing the numbers of CIN cases. Several novel immunoregulatory molecules have been discovered over the past few years, including the human leukocyte antigen G (HLA-G), which through interaction with its receptors exerts important tolerogenic functions. Several lines of evidence suggest that T-helper interleukin-17 (IL-17)-producing cells (Th17 cells) may play a role in antitumor immunity. However, recent reports have implicated Th17 cells and their cytokines in both pro and anti-tumorigenic processes. The aim of the study was to evaluate the roles of HLA-G and Th17 in the immunopathogenesis of CIN I. Analytical cross-sectional study with a control group using 58 cervical specimens from the files of a public university hospital providing tertiary-level care. We examined HLA-G and IL-17 expression in the cervical microenvironment by means of immunohistochemistry, and correlated these findings with clinical and pathological features. There was a greater tendency towards HLA-G and IL-17 expression in specimens that showed CIN I, thus suggesting that these molecules have a contribution towards cervical progression. These findings suggest that HLA-G and IL-17 expression may be an early marker for assessing the progression of cervical lesions.