EMAIL THIS PAGE TO A FRIEND
Kidney International
Primary osteoblast-like cells from patients with end-stage kidney disease reflect gene expression, proliferation, and mineralization characteristics ex vivo.
PMID 25354236