EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical Sciences
A prodrug approach involving in situ depot formation to achieve localized and sustained action of diclofenac after joint injection.
PMID 25354787