EMAIL THIS PAGE TO A FRIEND
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals
Unsaturated free fatty acids: a potential biomarker panel for early detection of gastric cancer.
PMID 25355065
Abstract
Changes in the levels of free fatty acids (FFAs) are closely associated with physiological status. Serum levels of C16:1, C18:3, C18:2, C18:1, C20:4, and C22:6 in 164 gastric cancer (GC) patients and 111 benign gastric disease (BGD) patients were significantly decreased compared with 252 healthy controls. Receiver operating characteristic analysis showed that the biomarker panel including C16:1, C18:3, C18:2, C20:4, and C22:6 presents a high diagnostic ability to differentiate early-stage GC patients from healthy controls plus BGD patients, with a sensitivity of 80.6% and a specificity of 72.7%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H5149
Heneicosanoic acid, analytical standard
C21H42O2