EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
New prenylated carbazole alkaloids from Zanthoxylum armatum.
PMID 25355272
Abstract
A phytochemical investigation on the ethyl acetate soluble fraction of Zanthoxylum armatum led in the isolation of two new prenylated alkaloids 2,6,7-trimethoxy-8-(3-methyl-2-butenyl)carbazole-3-carbaldehyde (1) and methyl-2,6,7-trimethoxy-8-(3-methyl-2-butenyl)carbazole-3-carboxylate (2), along with three known lignans cisamin (3), methyl pirpirtol (4), and fargesin (5) and one known alkaloid dictamine (6). Their structures were established on the basis of spectroscopic and crystallographic analysis and by comparison of the data with those in the literature. All the isolated compounds were screened for the DPPH free radical scavenging activity. Compounds 1, 2, and 6 showed profound activity while compounds 3-5 showed moderate activity.