EMAIL THIS PAGE TO A FRIEND
Journal of Pharmacology and Experimental Therapeutics
Pharmacologic targeting of sphingosine-1-phosphate receptor 1 improves the renal microcirculation during sepsis in the mouse.
PMID 25355645
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3944
SEW2871, ≥98% (HPLC), solid
C20H10F6N2OS