EMAIL THIS PAGE TO A FRIEND
Journal of Virology
Interactome analysis of the human respiratory syncytial virus RNA polymerase complex identifies protein chaperones as important cofactors that promote L-protein stability and RNA synthesis.
PMID 25355874