EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Biosynthesis of 8-hydroxyquinoline-2-carboxylic acid, an iron chelator from the gut of the lepidopteran Spodoptera littoralis.
PMID 25356857
Abstract
In the regurgitate (foregut content) of Spodoptera larvae we found high concentrations (0.5-5 mM) of 8-hydroxyquinoline-2-carboxylic acid (8-HQA). In a survey of different lepidopteran species, this compound was only detected in species belonging to the family of Noctuidae. 8-HQA was shown to derive from tryptophan metabolism. The amount of 8-HQA in the regurgitate was strongly dependent on the tryptophan content of the diet. In the insect 8-HQA is generated from tryptophan via kynurenine and 3-hydroxykynurenine. 8-HQA is produced by the larvae and not by their commensal gut bacteria. Analysis of different life stages of Spodoptera larvae revealed that 8-HQA is formed during the larval stage, probably acting as an iron chelator to control the gut microbiome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1771
3-Hydroxy-DL-kynurenine
C10H12N2O4
160660
Quinaldic acid, 98%
C10H7NO2
D120804
Xanthurenic acid, 96%
C10H7NO4