EMAIL THIS PAGE TO A FRIEND
European journal of gastroenterology & hepatology
Association of markers of bacterial translocation with immune activation in decompensated cirrhosis.
PMID 25357217
Abstract
Bacterial translocation (BT) may cause infections, in particular, spontaneous bacterial peritonitis (SBP). In the absence of overt infection, BT may further stimulate the immune system and contribute to haemodynamic alterations and complications. Bacterial DNA (bDNA) is claimed to be a promising surrogate marker for BT, although its clinical relevance has been questioned. In 38 cirrhotic patients with and without SBP, bDNA in blood and ascites were assessed by 16S rDNA quantitative PCR. Levels of lipopolysaccharide-binding protein in plasma and highly sensitive C-reactive protein, tumour necrosis factor-α, soluble urokinase plasminogen activating receptor, interleukin-6, interleukin 8, interferon-γ inducible protein-10 and vascular endothelial growth factor in plasma and ascites were measured by multiplex cytokine and ELISA assays. In patients without signs of SBP or positive cultures, we found a high frequency of bDNA but low concordance of bDNA between blood and ascites. Markers of inflammation were not significantly different between blood bDNA-positive (22%), ascites bDNA-positive (52%), and bDNA-negative patients. The 16S rDNA PCR failed to show bDNA in two out of six samples with SBP. Sequencing of positive samples did not determine the source of bDNA. bDNA as assessed by this PCR method was largely unrelated to markers of inflammation and does not seem to be of clinical value in the diagnosis of SBP. According to our results, bDNA is not a reliable marker of BT.