EMAIL THIS PAGE TO A FRIEND
Molecular reproduction and development
Inhibin alpha gene expression in human trophoblasts is regulated by interactions between TFAP2 and cAMP signaling pathways.
PMID 25358080
Abstract
Inhibin α (Inha) gene expression is regulated, in rat granulosa cells, via a cyclic 3',5'-adenosine monophosphate (AMP)-response element (CRE) found in a region of the promoter that is homologous to the human INHA promoter. We previously found that during in vitro cytotrophoblast differentiation, human INHA gene expression was regulated by TFAP2A via association with an AP-2 site located upstream of this CRE. The aim of this study was to evaluate if the human INHA gene was also regulated by cAMP in trophoblasts, and to investigate the possible crosstalk between TFAP2 and cAMP signaling pathways in the regulation of INHA gene expression. Treatment with cAMP or forskolin increased INHA mRNA expression by 7- and 2-fold in primary cytotrophoblasts and choriocarcinoma-derived BeWo cells, respectively. Treatment with the protein kinase A inhibitor H-89 reduced forskolin-induced luciferase activity by ∼40% in BeWo cells transfected with an INHA promoter-driven luciferase reporter vector. TFAP2 overexpression increased basal luciferase activity, whereas the dominant repressor KCREB abolished it. Surprisingly, mutation of the CRE also eliminated the TFAP2-induced transcription, although TFAP2 overexpression was still able to increase forskolin-induced luciferase activity when the AP-2 binding site, but not the CRE site, was mutated. Thus, INHA gene expression is upregulated by cAMP via CRE in human trophoblasts, and TFAP2 regulates this expression by interacting with CRE.