EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
High-resolution mass spectrometry associated with data mining tools for the detection of pollutants and chemical characterization of honey samples.
PMID 25358104
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0100000
Caffeine, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1085003
Caffeine, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1009
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
56396
Caffeine, certified reference material, TraceCERT®
C8H10N4O2
Y0000787
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1086006
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1095
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
41019
Melting point standard 235-237°C, analytical standard
C8H10N4O2
75035
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C8H10N4O2
295663
Propylene, ≥99%
C3H6
1613407
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
C2H3NaO2
613053
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
C2H3NaO2
306975
tert-Butyl methyl ether, anhydrous, 99.8%
C5H12O
675407
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
C5H12O
08603
tert-Butyl methyl ether, analytical standard
C5H12O
293210
tert-Butyl methyl ether, HPLC grade, for HPLC, 99.8%
C5H12O
PHR1560
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
C5H12O