Archives of pharmacal research
Organic cation transporter-mediated drug-drug interaction potential between berberine and metformin.
PMID 25359200
Berberine, the main active component of the herbal medicine Rhizoma Coptidis, has been reported to have hypoglycemic and insulin-sensitizing effects and, therefore, could be combined with metformin therapy. Thus, we assessed the potential drug-drug interactions between berberine and metformin. We investigated the in vitro inhibitory potency of berberine on metformin uptake in HEK293 cells overexpressing organic cation transporter (OCT) 1 and 2. To investigate whether this inhibitory effect of berberine on OCT1 and OCT2 could change the pharmacokinetics of metformin in vivo, we measured the effect of berberine co-administration on the pharmacokinetics of metformin at a single intravenous dose of 2xa0mg/kg metformin and 10xa0mg/kg berberine. In HEK293 cells, berberine inhibited OCT1- and OCT2-mediated metformin uptake in a concentration dependent manner and IC50 values for OCT1 and OCT2 were 7.28 and 11.3xa0μM, respectively. Co-administration of berberine increased the initial plasma concentration and AUC of metformin and decreased systemic clearance and volume of distribution of metformin in rats, suggesting that berberine inhibited disposition of metformin, which is governed by OCT1 and OCT2. Berberine inhibited the transport activity of OCT1 and OCT2 and showed significant potential drug-drug interactions with metformin in in vivo rats.