EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Structural basis for microRNA targeting.
PMID 25359968
Abstract
MicroRNAs (miRNAs) control expression of thousands of genes in plants and animals. miRNAs function by guiding Argonaute proteins to complementary sites in messenger RNAs (mRNAs) targeted for repression. We determined crystal structures of human Argonaute-2 (Ago2) bound to a defined guide RNA with and without target RNAs representing miRNA recognition sites. These structures suggest a stepwise mechanism, in which Ago2 primarily exposes guide nucleotides (nt) 2 to 5 for initial target pairing. Pairing to nt 2 to 5 promotes conformational changes that expose nt 2 to 8 and 13 to 16 for further target recognition. Interactions with the guide-target minor groove allow Ago2 to interrogate target RNAs in a sequence-independent manner, whereas an adenosine binding-pocket opposite guide nt 1 further facilitates target recognition. Spurious slicing of miRNA targets is avoided through an inhibitory coordination of one catalytic magnesium ion. These results explain the conserved nucleotide-pairing patterns in animal miRNA target sites first observed over two decades ago.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF86050199
Magnesium, microfoil, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Mg
GF56273777
Magnesium, microfoil, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Mg
GF80461964
Magnesium, microleaf, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF57746166
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF18270637
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF96495609
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF04987547
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF23637371
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF83427745
Magnesium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF86178189
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF83829178
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF61950618
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF23443646
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF54671924
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF28467636
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF72273157
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF78647293
Magnesium, wire reel, 1m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF43827071
Magnesium, wire reel, 1m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF30456053
Magnesium, wire reel, 1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF33990598
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF58454983
Magnesium, wire reel, 1m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF34143803
Magnesium, wire reel, 1m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF30911778
Magnesium, wire reel, 2m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF78384437
Magnesium, wire reel, 2m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF07006328
Magnesium, wire reel, 2m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF48824208
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF06872924
Magnesium, wire reel, 2m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF67975172
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Mg