EMAIL THIS PAGE TO A FRIEND
American journal of translational research
Maytansine-loaded star-shaped folate-core PLA-TPGS nanoparticles enhancing anticancer activity.
PMID 25360217
Abstract
The efficient delivery of therapeutic molecule agents into target cells of interest is a critical challenge to broad application of non-viral vector systems. In this research, maytansine-loaded star-shaped folate-core polylactide-D-α-tocopheryl polyethylene glycol 1000 succinate (FA-PLA-TPGS) block copolymer was applied to be a vector of maytansine for folate receptor positive (FR(+)) breast cancer therapy. The uptake of maytansine nanoparticles by SKBR3 cells were observed by fluorescence microscopy and confocal laser scanning microscopy. The cell viability of maytansine-NPs in SKBR3 cells was assessed according to the changed level of intracellular microtubules and apoptosis-associated proteins. The cytotoxicity of the SKBR3 cells was significantly increased by maytansine-NPs when compared with control groups. In conclusion, the maytansine-NPs offer a considerable potential formulation for FR-expressing tumor targeting biotherapy.