PloS one
LIN28B promotes colon cancer migration and recurrence.
PMID 25360631
Abstract
LIN28B is involved in "stemness" and tumourigenesis by negatively regulating the maturation of let-7 microRNA family members. In this study, we showed that LIN28B expression promotes migration and recurrence of colon cancer. Immunohistochemistry and reverse-transcription polymerase chain reactions were performed to detect LIN28B expression in colon cancer tissue microarrays, paraffin-embedded surgical resected tissues and cancer cells. Loss-of-function, migration and proliferation analyses were performed to delineate the potential roles of LIN28B in colon cancer. LIN28B was upregulated in colon cancer tissue compared to normal mucosa, and its overexpression correlated with reduced patient survival and increased tumour recurrence. LIN28B suppression inhibited the migration of SW480 colon cancer cells and facilitated the cytotoxicity induced by oxaliplatin in SW480 and HCT116 colon cancer cells. In conclusion, LIN28B overexpression contributes to colon tumourigenesis, and LIN28B may serve as a diagnostic tool and therapeutic target for colon cancer.