EMAIL THIS PAGE TO A FRIEND
The Biochemical Journal
Circular trimers of gelatinase B/matrix metalloproteinase-9 constitute a distinct population of functional enzyme molecules differentially regulated by tissue inhibitor of metalloproteinases-1.
PMID 25360794