EMAIL THIS PAGE TO A FRIEND
Molecular pharmaceutics
Dextrin-colistin conjugates as a model bioresponsive treatment for multidrug resistant bacterial infections.
PMID 25360900
Abstract
Polymer therapeutics offer potential benefits in the treatment of multidrug resistant (MDR) infections; affording targeted delivery of biologically active agents to the site of inflammation, potential decreases in systemic toxicity, and the retention of antimicrobial activity at the target site. As a prototype model, these studies developed and characterized a library of dextrin-colistin conjugates (dextrin molecular weight: 7500-48,000 g/mol) as a means of targeting the delivery of colistin. Optimum colistin release kinetics (following dextrin degradation by physiological concentrations of amylase (100 IU/L)) were observed in conjugates containing low molecular weight (∼7500 g/mol) dextrin with ∼1 mol % succinoylation (∼80% drug release within 48 h, compared to ∼33% from sodium colistin methanesulfonate (CMS, Colomycin)). These conjugates exhibited comparable antimicrobial activity to CMS in conventional MIC assays against a range of Gram-negative pathogens, but with significantly reduced in vitro toxicity toward kidney (IC₅₀ = CMS, 15.4 μg/mL; dextrin-colistin, 63.9 μg/mL) and macrophage (IC₅₀ = CMS, 111.3 μg/mL; dextrin-colistin, 303.9 μg/mL) cells. In vivo dose-escalation studies in rats demonstrated improved pharmacokinetics of the conjugates, with prolonged plasma levels of colistin (t₁/₂ 135-1271 min vs 53 min) and decreased toxicity, compared to colistin sulfate. These studies highlight the potential utility of "nanoantibiotic" polymer therapeutics to aid the safe, effective, and targeted delivery of colistin in the management of MDR infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
209198
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
CuO4S · 5H2O
C8027
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
CuO4S · 5H2O
C7631
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
CuO4S · 5H2O
61245
Copper(II) sulfate pentahydrate, purum p.a., crystallized, ≥99.0% (RT)
CuO4S · 5H2O
203165
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
CuO4S · 5H2O
469130
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
CuO4S · 5H2O
12849
Copper(II) sulfate pentahydrate, puriss., suitable for, meets analytical specification of Ph. Eur., BP, USP,FCC
CuO4S · 5H2O
31293
Copper(II) sulfate pentahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99-102%
CuO4S · 5H2O
C3036
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
CuO4S · 5H2O
RES10395-B7
Cupric sulfate pentahydrate, PharmaGrade, EP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
CuO4S · 5H2O
H17309
Hydrindantin, ≥97%
C18H10O6
L6883
Lithium acetate dihydrate, reagent grade
C2H3LiO2 · 2H2O
L4158
Lithium acetate dihydrate, BioXtra
C2H3LiO2 · 2H2O
62393
Lithium acetate dihydrate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
C2H3LiO2 · 2H2O
151173
Ninhydrin, ACS reagent
C9H6O4
N4876
Ninhydrin, suitable for amino acid detection
C9H6O4