EMAIL THIS PAGE TO A FRIEND
Aaps Pharmscitech
Influence of process and formulation parameters on dissolution and stability characteristics of Kollidon? VA 64 hot-melt extrudates.
PMID 25361900
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1667803
Alpha tocopheryl acid succinate, United States Pharmacopeia (USP) Reference Standard
C33H54O5
95255
D-α-Tocopherol succinate, BioXtra, ≥98.0% (HPLC)
C33H54O5
47782
D-α-Tocopherol succinate, analytical standard
C33H54O5
T3126
D-α-Tocopherol succinate, semisynthetic, 1210 IU/g
C33H54O5
P5835
Propyl 4-hydroxybenzoate, BioXtra
C10H12O3
54789
Propyl 4-hydroxybenzoate, tested according to Ph.Eur.
C10H12O3
P53357
Propyl 4-hydroxybenzoate, ≥99%
C10H12O3
W295101
Propyl 4-hydroxybenzoate, ≥99%
C10H12O3
P3650000
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C10H12O3
PHR1010
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C10H12O3
1577008
Propylparaben, United States Pharmacopeia (USP) Reference Standard
C10H12O3
T1610000
RRR-α-Tocopheryl hydrogen succinate, European Pharmacopoeia (EP) Reference Standard
C33H54O5
PHR1029
Tocopheryl Acid Succinate, a, Pharmaceutical Secondary Standard; Certified Reference Material
C33H54O5
14849
Triethyl citrate, analytical standard
C12H20O7
27500
Triethyl citrate, ≥98.0% (GC)
C12H20O7
W308307
Triethyl citrate, ≥99%, FCC, FG
C12H20O7
W308315
Triethyl citrate, natural, ≥97%, FG
C12H20O7
PHR1153
Triethyl citrate, Pharmaceutical Secondary Standard; Certified Reference Material
C12H20O7