EMAIL THIS PAGE TO A FRIEND
Journal of Proteome Research
Pathway and time-resolved benzo[a]pyrene toxicity on Hepa1c1c7 cells at toxic and subtoxic exposure.
PMID 25362887
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09689
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
C2H7NO2
73594
Ammonium acetate, for mass spectrometry, eluent additive for LC-MS
C2H7NO2
09691
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
C2H7NO2
78040
DL-Phenylalanine, puriss., ≥99.0% (NT)
C9H11NO2
W372609
DL-Phenylalanine, 99%, FCC
C9H11NO2
147966
DL-Phenylalanine, ReagentPlus®, 99%
C9H11NO2
T3300
DL-Tryptophan, ≥99% (HPLC)
C11H12N2O2
93680
DL-Tryptophan, puriss., ≥99.0% (NT)
C11H12N2O2
162698
DL-Tryptophan, ≥99% (HPLC)
C11H12N2O2
P1034
Phenyl isothiocyanate, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid
C7H5NS
78781
Phenyl isothiocyanate, for protein sequence analysis, ≥99.5% (GC)
C7H5NS
78780
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
C7H5NS
317861
Phenyl isothiocyanate, 99%, for protein sequencing
C7H5NS
139742
Phenyl isothiocyanate, reagent grade, 98%
C7H5NS
78787
Phenyl isothiocyanate solution, for protein sequence analysis, ~5% in heptane
C7H5NS
P1150000
Phenylalanine, European Pharmacopoeia (EP) Reference Standard
C9H11NO2
184527
Pyridine, spectrophotometric grade, ≥99%
C5H5N
02486
Pyridine, analytical standard
C5H5N
W296600
Pyridine, ≥99%
C5H5N
24-6510
Pyridine, SAJ first grade, ≥99.0%
C5H5N
PHR1558
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H5N
T2610000
Tryptophan, European Pharmacopoeia (EP) Reference Standard
C11H12N2O2
T2900000
Tyrosine, European Pharmacopoeia (EP) Reference Standard
C9H11NO3